Methyl 2-amino-4,6-difluorobenzoate 95%
Lieferant: Apollo Scientific
PC907296-1G
PC907296-250MG
APOSPC907296-1GEA
168
EUR
APOSPC907296-1G
APOSPC907296-250MG
Methyl 2-amino-4,6-difluorobenzoate 95%
Methyl 2-amino-4,6-difluorobenzoate
Formel:
C₈H₇F₂NO₂ Molecular Weight: 187,143 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12159819 CAS-Nummer: 379228-57-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...