1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine 95%
Lieferant: Apollo Scientific
Synonyme:
2-Methyl-5-trifluoromethyl-2H-pyrazol-3-ylamine
PC907339-1G
PC907339-250MG
PC907339-5G
APOSPC907339-1GEA
81.2
EUR
APOSPC907339-1G
APOSPC907339-250MG
APOSPC907339-5G
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine 95%
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-5-amine
Formel:
C₅H₆F₃N₃ Molecular Weight: 165.116 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06805773 CAS-Nummer: 149978-43-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...