Methyl 6-fluoro-5-methylpyridine-3-carboxylate 95%

Lieferant: Apollo Scientific

Synonyme: Methyl 6-fluoro-5-methylnicotinate

PC907364-1G PC907364-250MG PC907364-5G
APOSPC907364-1GEA 328 EUR
APOSPC907364-1G APOSPC907364-250MG APOSPC907364-5G
Methyl 6-fluoro-5-methylpyridine-3-carboxylate 95%
Methyl 6-fluoro-5-methylpyridine-3-carboxylate
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11616889
CAS-Nummer: 211122-38-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR