4-Acetyl-2-fluorobenzonitrile 97%
Lieferant: Apollo Scientific
PC907486-1G
APOSPC907486-1GEA
95.7
EUR
APOSPC907486-1G
4-Acetyl-2-fluorobenzonitrile 97%
4-Acetyl-2-fluorobenzonitrile
Formel:
C₉H₆FNO Molecular Weight: 163,148 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10575008 CAS-Nummer: 214760-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...