4-Fluorophenyl methanesulphonate 98%

Lieferant: Apollo Scientific

PC907562-1G PC907562-250MG
APOSPC907562-1GEA 886 EUR
APOSPC907562-1G APOSPC907562-250MG
4-Fluorophenyl methanesulphonate 98%
4-Fluorophenyl methanesulphonate
Formel: C₇H₇FO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD16036342
CAS-Nummer: 72358-72-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR