7-Fluoro-1,2,3,4-tetrahydroisoquinoline 95%

Lieferant: Apollo Scientific

PC907573-1G PC907573-250MG
APOSPC907573-1GEA 790 EUR
APOSPC907573-1G APOSPC907573-250MG
7-Fluoro-1,2,3,4-tetrahydroisoquinoline 95%
7-Fluoro-1,2,3,4-tetrahydroisoquinoline
Formel: C₉H₁₀FN
Molecular Weight: 151.181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094746
CAS-Nummer: 406923-91-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR