1-(3,4-Difluorophenyl)cyclopropanamine 98%

Lieferant: Apollo Scientific

PC907867-100MG PC907867-1G PC907867-250MG
APOSPC907867-100MGEA 89.9 EUR
APOSPC907867-100MG APOSPC907867-1G APOSPC907867-250MG
1-(3,4-Difluorophenyl)cyclopropanamine 98%
1-(3,4-Difluorophenyl)cyclopropanamine
Formel: C₉H₉F₂N
Molecular Weight: 169.171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09910057
CAS-Nummer: 474709-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR