4-Fluoro-N-methyl-3-(trifluoromethyl)aniline 95%

Lieferant: Apollo Scientific

PC908002-1G PC908002-5G
APOSPC908002-1GEA 203 EUR
APOSPC908002-1G APOSPC908002-5G
4-Fluoro-N-methyl-3-(trifluoromethyl)aniline 95%
4-Fluoro-N-methyl-3-(trifluoromethyl)aniline
Formel: C₈H₇F₄N
Molecular Weight: 193,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11655738
CAS-Nummer: 216982-01-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR