3-Amino-2,4-difluorobenzonitrile 95%

Lieferant: Apollo Scientific

PC908025-1G PC908025-5G
APOSPC908025-1GEA 190 EUR
APOSPC908025-1G APOSPC908025-5G
3-Amino-2,4-difluorobenzonitrile 95%
3-Amino-2,4-difluorobenzonitrile
Formel: C₇H₄F₂N₂
Molecular Weight: 154,117 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26687812
CAS-Nummer: 1505597-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR