3-(Difluorohydroxy)-4-hydroxybenzonitrile 95%
Lieferant: Apollo Scientific
PC908034-1G
PC908034-5G
APOSPC908034-1GEA
156
EUR
APOSPC908034-1G
APOSPC908034-5G
3-(Difluorohydroxy)-4-hydroxybenzonitrile 95%
3-(Difluorohydroxy)-4-hydroxybenzonitrile
Formel:
C₈H₅F₂NO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393963 CAS-Nummer: 1261823-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...