2,5-Difluoro-4-formylbenzonitrile 95%

Lieferant: Apollo Scientific

PC908037-1G PC908037-250MG
APOSPC908037-1GEA 387 EUR
APOSPC908037-1G APOSPC908037-250MG
2,5-Difluoro-4-formylbenzonitrile 95%
2,5-Difluoro-4-formylbenzonitrile
Formel: C₈H₃F₂NO
Molecular Weight: 167.112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16876871
CAS-Nummer: 433940-02-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR