5-Bromo-2,4-difluoroiodobenzene 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Bromo-2,4-difluoro-5-iodobenzene

PC9081-1G PC9081-25G PC9081-5G
APOSPC9081-1GEA 21.9 EUR
APOSPC9081-1G APOSPC9081-25G APOSPC9081-5G
5-Bromo-2,4-difluoroiodobenzene 98%
5-Bromo-2,4-difluoroiodobenzene
Formel: C₆H₂BrF₂I
Molecular Weight: 318,885 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08458121
CAS-Nummer: 914636-91-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR