5-(Trifluoromethyl)thiophene-2-carboxylic acid 95%
Lieferant: Apollo Scientific
PC908138-1G
PC908138-250MG
APOSPC908138-1GEA
230
EUR
APOSPC908138-1G
APOSPC908138-250MG
5-(Trifluoromethyl)thiophene-2-carboxylic acid 95%
5-(Trifluoromethyl)thiophene-2-carboxylic acid
Formel:
C₆H₃F₃O₂S Molecular Weight: 196.147 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04421350 CAS-Nummer: 128009-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...