5,8-Difluorochroman-4-one 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC908206-100MG
PC908206-250MG
APOSPC908206-100MGEA
36.4
EUR
APOSPC908206-100MG
APOSPC908206-250MG
5,8-Difluorochroman-4-one 97%
5,8-Difluorochroman-4-one
Formel:
C₉H₆F₂O₂ Molecular Weight: 184,14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11518475 CAS-Nummer: 942196-01-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...