4-Fluoroisoquinoline-5-sulphonyl chloride 95%

Lieferant: Apollo Scientific

PC908211-10G PC908211-1G PC908211-5G
APOSPC908211-10GEA 1290 EUR
APOSPC908211-10G APOSPC908211-1G APOSPC908211-5G
4-Fluoroisoquinoline-5-sulphonyl chloride 95%
4-Fluoroisoquinoline-5-sulphonyl chloride
Formel: C₉H₅ClFNO₂S
Molecular Weight: 245.658 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19200505
CAS-Nummer: 194032-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR