4-Chloro-6-fluoro-1H-indole-2,3-dione 90%

Lieferant: Apollo Scientific

Synonyme: 4-Chloro-6-fluoroindoline-2,3-dione

PC908426-1G PC908426-250MG
APOSPC908426-1GEA 264 EUR
APOSPC908426-1G APOSPC908426-250MG
4-Chloro-6-fluoro-1H-indole-2,3-dione 90%
4-Chloro-6-fluoro-1H-indole-2,3-dione
Formel: C₈H₃ClFNO₂
Molecular Weight: 199.566 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20485537
CAS-Nummer: 940054-45-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR