7-Bromo-5-fluoroindoline-2,3-dione 98%

Lieferant: Apollo Scientific
Achtung

PC908520-1G PC908520-250MG PC908520-5G
APOSPC908520-1GEA 91.5 EUR
APOSPC908520-1G APOSPC908520-250MG APOSPC908520-5G
7-Bromo-5-fluoroindoline-2,3-dione 98%
7-Bromo-5-fluoroindoline-2,3-dione
Formel: C₈H₃BrFNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03150727
CAS-Nummer: 380431-78-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR