6-Fluoronaphthalen-1-ol 95+%

Lieferant: Apollo Scientific
Achtung

PC908525-100MG PC908525-250MG
APOSPC908525-100MGEA 84.1 EUR
APOSPC908525-100MG APOSPC908525-250MG
6-Fluoronaphthalen-1-ol 95+%
6-Fluoronaphthalen-1-ol
Formel: C₁₀H₇FO
Molecular Weight: 162,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15528810
CAS-Nummer: 804498-72-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR