2,4-Difluoro-N-methyl-benzylamine 97%
Lieferant: Apollo Scientific
Synonyme:
1-(2,4-Difluorophenyl)-N-methylmethanamine
PC908597-1G
APOSPC908597-1GEA
127
EUR
APOSPC908597-1G
2,4-Difluoro-N-methyl-benzylamine 97%
2,4-Difluoro-N-methyl-benzylamine
Formel:
C₈H₉F₂N Molecular Weight: 157,161 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09047006 CAS-Nummer: 696589-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...