2,2,2-Trifluoro-1-(furan-2-yl)ethanone 95%

Lieferant: Apollo Scientific

PC908598-1G PC908598-250MG
APOSPC908598-1GEA 244 EUR
APOSPC908598-1G APOSPC908598-250MG
2,2,2-Trifluoro-1-(furan-2-yl)ethanone 95%
2,2,2-Trifluoro-1-(furan-2-yl)ethanone
Formel: C₆H₃F₃O₂
Molecular Weight: 164,082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07380746
CAS-Nummer: 18207-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR