3-(2,6-Difluorophenyl)propanoic acid 98%
Lieferant: Apollo Scientific
PC908697-1G
PC908697-5G
APOSPC908697-1GEA
270
EUR
APOSPC908697-1G
APOSPC908697-5G
3-(2,6-Difluorophenyl)propanoic acid 98%
3-(2,6-Difluorophenyl)propanoic acid
Formel:
C₉H₈F₂O₂ Molecular Weight: 186.155 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06823978 CAS-Nummer: 167683-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...