1-Fluoro-2,4-dimethyl-5-nitrobenzene 95%

Lieferant: Apollo Scientific

PC908761-1G PC908761-250MG
APOSPC908761-1GEA 190 EUR
APOSPC908761-1G APOSPC908761-250MG
1-Fluoro-2,4-dimethyl-5-nitrobenzene 95%
1-Fluoro-2,4-dimethyl-5-nitrobenzene
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12545823
CAS-Nummer: 345-22-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR