6-Chloro-5-fluoro-2-methylpyrimidin-4-ol 97%

Lieferant: Apollo Scientific

PC908766-1G PC908766-250MG
APOSPC908766-1GEA 85.7 EUR
APOSPC908766-1G APOSPC908766-250MG
6-Chloro-5-fluoro-2-methylpyrimidin-4-ol 97%
6-Chloro-5-fluoro-2-methylpyrimidin-4-ol
Formel: C₅H₄ClFN₂O
Molecular Weight: 162.549 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23099034
CAS-Nummer: 105806-14-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR