4-Fluoro-2-(methylsulphonyl)aniline 95%

Lieferant: Apollo Scientific

PC908779-1G PC908779-5G
APOSPC908779-1GEA 127 EUR
APOSPC908779-1G APOSPC908779-5G
4-Fluoro-2-(methylsulphonyl)aniline 95%
4-Fluoro-2-(methylsulphonyl)aniline
Formel: C₇H₈FNO₂S
Molecular Weight: 189,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13191582
CAS-Nummer: 1197193-21-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR