N-(2,3-Difluorobenzyl)-N-methylamine 95%

Lieferant: Apollo Scientific

PC908829-1G PC908829-5G
APOSPC908829-1GEA 284 EUR
APOSPC908829-1G APOSPC908829-5G
N-(2,3-Difluorobenzyl)-N-methylamine 95%
N-(2,3-Difluorobenzyl)-N-methylamine
Formel: C₈H₉F₂N
Molecular Weight: 157,161 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12147620
CAS-Nummer: 906645-41-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR