(S)-2-Amino-2-(3-fluorophenyl)ethanol 97%

Lieferant: Apollo Scientific

PC908892-1G PC908892-250MG
APOSPC908892-1GEA 127 EUR
APOSPC908892-1G APOSPC908892-250MG
(S)-2-Amino-2-(3-fluorophenyl)ethanol 97%
(S)-2-Amino-2-(3-fluorophenyl)ethanol
Formel: C₈H₁₀FNO
Molecular Weight: 155,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09256370
CAS-Nummer: 325152-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR