Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium trifluoro-[[4-[(2-methylpropan-2-yl)oxycarbonyl]piperazin-1-yl]methyl]boranuide

PC909040-1G PC909040-5G
APOSPC909040-1GEA 29 EUR
APOSPC909040-1G APOSPC909040-5G
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected 95%
Potassium (piperazin-1-yl)methyltrifluoroborate, N-4 BOC protected
Formel: C₁₀H₁₉BF₃N₂O₂·K
Lagertemperatur: Raumtemperatur
MDL: MFCD10566514
CAS-Nummer: 936329-97-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR