5-Fluoro-N-methyl-2-nitroaniline 97%

Lieferant: Apollo Scientific

PC909138-1G PC909138-5G
APOSPC909138-1GEA 134 EUR
APOSPC909138-1G APOSPC909138-5G
5-Fluoro-N-methyl-2-nitroaniline 97%
5-Fluoro-N-methyl-2-nitroaniline
Formel: C₇H₇FN₂O₂
Molecular Weight: 170,141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16744213
CAS-Nummer: 120381-42-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR