8-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%
Lieferant: Apollo Scientific
Synonyme:
8-Fluoro-4,4-dimethyl-3,4-dihydroquinolin-2(1H)-one
PC909145-25G
PC909145-5G
APOSPC909145-25GEA
836
EUR
APOSPC909145-25G
APOSPC909145-5G
8-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one 98%
8-Fluoro-4,4-dimethyl-1,3-dihydroquinolin-2-one
Formel:
C₁₁H₁₂FNO Molecular Weight: 193.217 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12913841 CAS-Nummer: 1187933-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...