1,4-Difluoro-2-(prop-2-yn-1-yloxy)benzene 95%

Lieferant: Apollo Scientific

PC909158-1G PC909158-250MG
APOSPC909158-1GEA 190 EUR
APOSPC909158-1G APOSPC909158-250MG
1,4-Difluoro-2-(prop-2-yn-1-yloxy)benzene 95%
1,4-Difluoro-2-(prop-2-yn-1-yloxy)benzene
Formel: C₉H₆F₂O
Molecular Weight: 168,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18207073
CAS-Nummer: 457628-36-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR