2-(4-Fluorophenyl)ethanethioamide 98%

Lieferant: Apollo Scientific

PC909166-10G PC909166-1G PC909166-5G
APOSPC909166-10GEA 502 EUR
APOSPC909166-10G APOSPC909166-1G APOSPC909166-5G
2-(4-Fluorophenyl)ethanethioamide 98%
2-(4-Fluorophenyl)ethanethioamide
Formel: C₈H₈FNS
Lagertemperatur: Raumtemperatur
MDL: MFCD09814508
CAS-Nummer: 351-82-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR