6-(4-Fluorophenyl)-2-hydroxypyridine 98%

Lieferant: Apollo Scientific

PC909188-1G PC909188-5G
APOSPC909188-1GEA 145 EUR
APOSPC909188-1G APOSPC909188-5G
6-(4-Fluorophenyl)-2-hydroxypyridine 98%
6-(4-Fluorophenyl)-2-hydroxypyridine
Formel: C₁₁H₈FNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11876187
CAS-Nummer: 1111111-04-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR