3-(4-Fluorophenyl)phenol 98%
Lieferant: Apollo Scientific
PC909236-1G
PC909236-5G
APOSPC909236-1GEA
0
EUR
APOSPC909236-1G
APOSPC909236-5G
3-(4-Fluorophenyl)phenol 98%
3-(4-Fluorophenyl)phenol
Formel:
C₁₂H₉FO Lagertemperatur: Raumtemperatur |
MDL:
MFCD06802305 CAS-Nummer: 10540-41-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...