(R)-2-(3-Fluorophenyl)pyrrolidine 95%

Lieferant: Apollo Scientific

PC909317-1G PC909317-250MG
APOSPC909317-1GEA 272 EUR
APOSPC909317-1G APOSPC909317-250MG
(R)-2-(3-Fluorophenyl)pyrrolidine 95%
(R)-2-(3-Fluorophenyl)pyrrolidine
Formel: C₁₀H₁₂FN
Molecular Weight: 165,207 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06762528
CAS-Nummer: 920274-03-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR