6-(4-Trifluoromethoxyphenyl)picolinic acid 95%

Lieferant: Apollo Scientific

Synonyme: 6-[4-(Trifluoromethoxy)phenyl]pyridine-2-carboxylic acid

PC909383-1G PC909383-250MG
APOSPC909383-1GEA 491 EUR
APOSPC909383-1G APOSPC909383-250MG
6-(4-Trifluoromethoxyphenyl)picolinic acid 95%
6-(4-Trifluoromethoxyphenyl)picolinic acid
Formel: C₁₃H₈F₃NO₃
Molecular Weight: 283,203 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06410074
CAS-Nummer: 887982-16-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR