5-Bromo-7-fluorobenzofuran
Lieferant: Apollo Scientific
|
Achtung
|
PC909455-1G
PC909455-250MG
APOSPC909455-1GEA
277
EUR
APOSPC909455-1G
APOSPC909455-250MG
5-Bromo-7-fluorobenzofuran
5-Bromo-7-fluorobenzofuran
Formel:
C₈H₄BrFO Molecular Weight: 215.019 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09056787 CAS-Nummer: 286836-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...