5-(4-Fluorophenyl)pyridin-2-amine 98%

Lieferant: Apollo Scientific

PC909623-1G PC909623-5G
APOSPC909623-1GEA 190 EUR
APOSPC909623-1G APOSPC909623-5G
5-(4-Fluorophenyl)pyridin-2-amine 98%
5-(4-Fluorophenyl)pyridin-2-amine
Formel: C₁₁H₉FN₂
Molecular Weight: 188,201 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802133
CAS-Nummer: 503536-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR