7,8-Difluorochroman-4-one 97%

Lieferant: Apollo Scientific
Achtung

PC909653-1G PC909653-250MG PC909653-5G
APOSPC909653-1GEA 130 EUR
APOSPC909653-1G APOSPC909653-250MG APOSPC909653-5G
7,8-Difluorochroman-4-one 97%
7,8-Difluorochroman-4-one
Formel: C₉H₆F₂O₂
Molecular Weight: 184,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11518476
CAS-Nummer: 890840-90-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR