5-Fluoro-1,3-benzoxazol-2-amine 96%

Lieferant: Apollo Scientific

Synonyme: 5-Fluorobenzo[d]oxazol-2-amine

PC909834-1G
APOSPC909834-1GEA 112 EUR
APOSPC909834-1G
5-Fluoro-1,3-benzoxazol-2-amine 96%
5-Fluoro-1,3-benzoxazol-2-amine
Formel: C₇H₅FN₂O
Molecular Weight: 152,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01664142
CAS-Nummer: 1682-39-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR