1-Fluoro-4-nitronaphthalene 95%

Lieferant: Apollo Scientific

PC909909-100MG PC909909-250MG
APOSPC909909-100MGEA 219 EUR
APOSPC909909-100MG APOSPC909909-250MG
1-Fluoro-4-nitronaphthalene 95%
1-Fluoro-4-nitronaphthalene
Formel: C₁₀H₆FNO₂
Molecular Weight: 191,159 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00156398
CAS-Nummer: 341-92-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR