2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene 95%

Lieferant: Apollo Scientific

PC910138-1G PC910138-250MG PC910138-5G PC910138-25G
APOSPC910138-1GEA 393 EUR
APOSPC910138-1G APOSPC910138-250MG APOSPC910138-5G APOSPC910138-25G
2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene 95%
2-Chloro-1-fluoro-4-(trifluoromethoxy)benzene
Formel: C₇H₃ClF₄O
Molecular Weight: 214.545 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22580940
CAS-Nummer: 128540-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR