2-Chloro-5-fluoropyridin-4-ol 96%
Lieferant: Apollo Scientific
Synonyme:
2-Chloro-5-fluoro-pyridin-4-ol
PC910219-1G
PC910219-5G
APOSPC910219-1GEA
74.1
EUR
APOSPC910219-1G
APOSPC910219-5G
2-Chloro-5-fluoropyridin-4-ol 96%
2-Chloro-5-fluoropyridin-4-ol
Formel:
C₅H₃ClFNO Molecular Weight: 147.535 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13189882 CAS-Nummer: 1196153-96-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...