7-Fluoro-1-methyl-1H-indole-2,3-dione 95%

Lieferant: Apollo Scientific

Synonyme: 7-Fluoro-1-methylindoline-2,3-dione

PC910563-5G
APOSPC910563-5GEA 82.8 EUR
APOSPC910563-5G
7-Fluoro-1-methyl-1H-indole-2,3-dione 95%
7-Fluoro-1-methyl-1H-indole-2,3-dione
Formel: C₉H₆FNO₂
Molecular Weight: 179,148 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21984607
CAS-Nummer: 875003-43-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR