7-Fluoronaphthalen-1-ol 97%

Lieferant: Apollo Scientific
Achtung

PC910642-100MG PC910642-250MG
APOSPC910642-100MGEA 52.2 EUR
APOSPC910642-100MG APOSPC910642-250MG
7-Fluoronaphthalen-1-ol 97%
7-Fluoronaphthalen-1-ol
Formel: C₁₀H₇FO
Molecular Weight: 162,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00077540
CAS-Nummer: 3132-92-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR