5-Fluoro-2,4-dihydro-1,4-benzoxazin-3-one 98%

Lieferant: Apollo Scientific

PC910686-1G PC910686-250MG
APOSPC910686-1GEA 255 EUR
APOSPC910686-1G APOSPC910686-250MG
5-Fluoro-2,4-dihydro-1,4-benzoxazin-3-one 98%
5-Fluoro-2,4-dihydro-1,4-benzoxazin-3-one
Formel: C₈H₆FNO₂
Molecular Weight: 167.137 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20441929
CAS-Nummer: 1221502-66-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR