6-Fluoro-3-methylisoquinoline 98%

Lieferant: Apollo Scientific

PC910719-1G PC910719-250MG
APOSPC910719-1GEA 0 EUR
APOSPC910719-1G APOSPC910719-250MG
6-Fluoro-3-methylisoquinoline 98%
6-Fluoro-3-methylisoquinoline
Formel: C₁₀H₈FN
Lagertemperatur: Raumtemperatur
MDL: MFCD22571325
CAS-Nummer: 1221445-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR