2-[(2-Fluoropyridin-3-yl)methylidene]malononitrile 96%

Lieferant: Apollo Scientific

PC910738-1G PC910738-5G
APOSPC910738-1GEA 219 EUR
APOSPC910738-1G APOSPC910738-5G
2-[(2-Fluoropyridin-3-yl)methylidene]malononitrile 96%
2-[(2-Fluoropyridin-3-yl)methylidene]malononitrile
Formel: C₉H₄FN₃
Molecular Weight: 173.147 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26383424
CAS-Nummer: 1820674-40-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR