7-Fluoro-1,3-dihydroquinazoline-2,4-dione 96%

Lieferant: Apollo Scientific

Synonyme: 7-Fluoroquinazoline-2,4(1H,3H)-dione

PC910746-1G PC910746-5G
APOSPC910746-1GEA 36.4 EUR
APOSPC910746-1G APOSPC910746-5G
7-Fluoro-1,3-dihydroquinazoline-2,4-dione 96%
7-Fluoro-1,3-dihydroquinazoline-2,4-dione
Formel: C₈H₅FN₂O₂
Molecular Weight: 180,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09954843
CAS-Nummer: 76088-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR