3-(2-Fluoropyridin-3-yl)propanoic acid 96%

Lieferant: Apollo Scientific

PC910748-1G PC910748-250MG
APOSPC910748-1GEA 632 EUR
APOSPC910748-1G APOSPC910748-250MG
3-(2-Fluoropyridin-3-yl)propanoic acid 96%
3-(2-Fluoropyridin-3-yl)propanoic acid
Formel: C₈H₈FNO₂
Molecular Weight: 169,153 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18803197
CAS-Nummer: 944998-14-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR