1,1,1-Trifluoro-3-(pyridin-2-yl)propan-2-amine 98%

Lieferant: Apollo Scientific

PC910753-1G PC910753-250MG
APOSPC910753-1GEA 806 EUR
APOSPC910753-1G APOSPC910753-250MG
1,1,1-Trifluoro-3-(pyridin-2-yl)propan-2-amine 98%
1,1,1-Trifluoro-3-(pyridin-2-yl)propan-2-amine
Formel: C₈H₉F₃N₂
Molecular Weight: 190,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11178764
CAS-Nummer: 1094372-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR